missing translation for 'onlineSavingsMsg'
Learn More

Durable™ Desk mat, durable, 400mm x 530mm, clear/black

Product Code. 15414234
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 15414234 Supplier Durable™ Supplier No. 1510004
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Features clear overlay to keep telephone numbers, notes and reminders close at hand. Non-slip foam backing.
  • Clear/black
  • Size - 400mm x 530mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.