missing translation for 'onlineSavingsMsg'
Learn More

Desipramine hydrochloride, MedChemExpress

Product Code. 16422266
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422266 500 mg 500mg
16418716 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16422266 Supplier Medchem Express Supplier No. HYB1272
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Desipramine hydrochloride is an inhibitor of norepinephrine transporter (NET), 5-HT transporter (SERT) and dopamine transporter (DAT) with Kis of 4, 61 and 78,720 nM, respectively.

Desipramine hydrochloride is an inhibitor of norepinephrine transporter (NET), 5-HT transporter (SERT) and dopamine transporter (DAT) with Kis of 4, 61 and 78,720 nM, respectively.

Specifications

Chemical Name or Material Desipramine hydrochloride
CAS 58-28-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H23ClN2
Quantity 500 mg
Solubility Information DMSO : ≥ 100 mg/mL (330.21 mM) ∣H2O : 100 mg/mL (330.21 mM; Need ultrasonic)
SMILES CNCCCN1C2=CC=CC=C2CCC3=CC=CC=C31.[H]Cl
Molecular Weight (g/mol) 302.84
Formula Weight 302.84
Percent Purity 99.64%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.