missing translation for 'onlineSavingsMsg'
Learn More

Desipramine.HCl, Lipomed

Product Code. 30462560
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462560 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462560 Supplier Lipomed Supplier No. LPMDSP1093HC10
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Desipramine.HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desipramine Hydrochloride
CAS 58-28-6
Analyte or Component Names Desipramine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C18 H22 N2 . Cl H
Synonym Imipramine Hydrochloride Imp. A (EP), Imipramine Imp. A (EP), Desipramine Hydrochloride, 3-(10,11-Dihydro-5 H-dibenzo[b,f]azepin-5-yl)-N-methylpropan-1-amine Hydrochloride, Imipramine Hydrochloride Imp. A (EP) as Hydrochloride, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A, RMI 9384 A
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.CNCCCN1c2ccccc2CCc3ccccc13
IUPAC Name 3-(5,6-dihydrobenzo[b][1 ]benzazepin-11-yl)-N-methylpropan-1-amine;hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 302.84
Quantity 10 mg
Formula Weight 302.155 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.