missing translation for 'onlineSavingsMsg'
Learn More

Desipramine-d3, TRC

Product Code. 30362273
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30362273 25 mg 25mg
30356818 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30362273 Supplier Toronto Research Chemicals Supplier No. TRCD29005325MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65100-49-4
Molecular Formula C18 2H3 H20 N3
Molecular Weight (g/mol) 284.41
Synonym Desipramine D3
IUPAC Name N,N-dimethyl-3-(2,3,4-trideuterio-5,6-dihydrobenzo[b][1,4]benzodiazepin-11-yl)propan-1-amine
SMILES [2H]c1cc2N(CCCN(C)C)c3ccccc3CNc2c([2H])c1[2H]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H23N3/c1-20(2)12-7-13-21-17-10-5-3-8-15(17)14-19-16-9-4-6-11-18(16)21/h3-6,8-11,19H,7,12-14H2,1-2H3/i4D,6D,9D
Formula Weight 284.208
Percent Purity >95%
Chemical Name or Material Desipramine Methyl-d3
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.