missing translation for 'onlineSavingsMsg'
Learn More

Deshydroxy Sotalol Hydrochloride, TRC

Product Code. 30363144
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30363144 10 mg 10mg
30361233 50 mg 50mg
30360239 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363144 Supplier Toronto Research Chemicals Supplier No. TRCD29005510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16974-44-0
Molecular Formula C12 H20 N2 O2 S . Cl H
Molecular Weight (g/mol) 292.83
Synonym N-[4-[2-[(1-Methylethyl)amino]ethyl]phenyl]-methanesulphonamide Hydrochloride, Sotalol Hydrochloride Imp. A (EP) as Hydrochloride
IUPAC Name N-[4-[2-(propan-2-ylamino)ethyl]phenyl]methanesulfonamide;hydrochloride
SMILES Cl.CC(C)NCCc1ccc(NS(=O)(=O)C)cc1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H20N2O2S.ClH/c1-10(2)13-9-8-11-4-6-12(7-5-11)14-17(3,15)16;/h4-7,10,13-14H,8-9H2,1-3H3;1H
Formula Weight 292.1012
Chemical Name or Material N-[4-[2-[(1-Methylethyl)amino]ethyl]phenyl]-methanesulphonamide Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.