missing translation for 'onlineSavingsMsg'
Learn More

Desfuroyl Ceftiofur (>85%), TRC

Product Code. 17902345
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17902345 2.5 mg 2.5mg
17991355 10 mg 10mg
17931155 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17902345 Supplier Toronto Research Chemicals Supplier No. TRCD2899802.5MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120882-22-6
Molecular Formula C14 H15 N5 O5 S3
Molecular Weight (g/mol) 429.49
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-3-(mercaptomethyl)-8-oxo-, (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-(mercaptomethyl)-8-oxo-, [6R-[6α,7β(Z)]]-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-(mercaptomethyl)-8-oxo-, (6R,7R)- (9CI), (6R,7R)-7-[[(2Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-3-(mercaptomethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, Defuroylceftiofur, Desfuroylceftiofur, Desfuroyl Ceftiofur
IUPAC Name (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-3-(sulfanylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)CS)\c3csc(N)n3

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C14H15N5O5S3/c1-24-18-7(6-4-27-14(15)16-6)10(20)17-8-11(21)19-9(13(22)23)5(2-25)3-26-12(8)19/h4,8,12,25H,2-3H2,1H3,(H2,15,16)(H,17,20)(H,22,23)/b18-7-/t8-,12-/m1/s1
Formula Weight 429.0235
Chemical Name or Material Desfuroyl Ceftiofur
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.