missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desethylamiodarone Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Specifications
Specifications
| Chemical Name or Material | N-Desethylamiodarone HCl |
| CAS | 96027-74-6 |
| Analyte or Component Names | N-Desethylamiodarone HCl |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H25 I2 N O3 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | (2-Butylbenzofuran-3-yl)[4-[2-(ethylamino)ethoxy]-3,5-diiodophenyl]methanone Hydrochloride (Desethylamiodarone Hydrochloride), Amiodarone Hydrochloride Imp. B (EP), Amiodarone Imp. B (EP), (2-Butylbenzofuran-3-yl)[4-[2-(ethylamino)ethoxy]-3,5-diiodophenyl]methanone Hydrochloride, Amiodarone Hydrochloride Imp. B (EP) as Hydrochloride, Desethylamiodarone Hydrochloride, Amiodarone Hydrochloride Impurity B as Hydrochloride, Amiodarone Impurity B as Hydrochloride |
| InChI Formula | InChI=1S/C23H25I2NO3.ClH/c1-3-5-9-20-21(16-8-6-7-10-19(16)29-20)22(27)15-13-17(24)23(18(25)14-15)28-12-11-26-4-2;/h6-8,10,13-14,26H,3-5,9,11-12H2,1-2H3;1H |
| Solution Type | Single Solution |
| SMILES | Cl.CCCCc1oc2ccccc2c1C(=O)c3cc(I)c(OCCNCC)c(I)c3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?