missing translation for 'onlineSavingsMsg'
Learn More

Desert Hedgehog/Dhh Antibody (2D5), Novus Biologicals™

Product Code. 18377909 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377909 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377909 Supplier Novus Biologicals Supplier No. H00050846M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Desert Hedgehog/Dhh Monoclonal antibody specifically detects Desert Hedgehog/Dhh in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desert Hedgehog/Dhh
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2D5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_066382
Gene Alias desert hedgehog, desert hedgehog (Drosophila) homolog, desert hedgehog homolog, desert hedgehog protein, HHG-3MGC35145
Host Species Mouse
Immunogen DHH (NP_066382, 210 a.a. ~ 310 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SGERKGLRELHRGDWVLAADASGRVVPTPVLLFLDRDLQRRASFVAVETEWPPRKLLLTPWHLVFAARGPAPAPGDFAPVFARRLRAGDSVLAPGGDALRP*
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 50846
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.