missing translation for 'onlineSavingsMsg'
Learn More

Descyano Nirmatrelvir Acetamide, TRC

Product Code. 30382677
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382677 1 g
30387887 100 mg
30372133 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382677 Supplier Toronto Research Chemicals Supplier No. TRCD7362251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2755812-81-6
Molecular Formula C23 H34 F3 N5 O5
Molecular Weight (g/mol) 517.542
Synonym (1R,2S,5S)-N-[(2S)-1-Amino-1-Oxo-3-((3S)-2-Oxopyrrolidin-3-Yl)Propan-2-Yl]-6,6-Dimethyl-3-[3-Methyl-N-(Trifluoroacetyl)-L-Valyl]-3-Azabicyclo[3.1.0]Hexane-2-Carboxamide
IUPAC Name (1R,2S,5S)-N-[(2S)-1-amino-1-oxo-3-[(3S)-2-oxopyrrolidin-3-yl]propan-2-yl]-3-[(2S)-3,3-dimethyl-2-[(2,2,2-trifluoroacetyl)amino]butanoyl]-6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxamide
SMILES CC(C)(C)[C@H](NC(=O)C(F)(F)F)C(=O)N1C[C@H]2[C@@H]([C@H]1C(=O)N[C@@H](C[C@@H]3CCNC3=O)C(=O)N)C2(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C23H34F3N5O5/c1-21(2,3)15(30-20(36)23(24,25)26)19(35)31-9-11-13(22(11,4)5)14(31)18(34)29-12(16(27)32)8-10-6-7-28-17(10)33/h10-15H,6-9H2,1-5H3,(H2,27,32)(H,28,33)(H,29,34)(H,30,36)/t10-,11-,12-,13-,14-,15+/m0/s1
Formula Weight 517.251
Chemical Name or Material Descyano Nirmatrelvir Acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.