missing translation for 'onlineSavingsMsg'
Learn More

Descyano Nirmatrelvir Acetamide, TRC

Product Code. 30387887
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387887 100 mg
30372133 250 mg
30382677 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387887 Supplier Toronto Research Chemicals Supplier No. TRCD736225100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2755812-81-6
Molecular Formula C23 H34 F3 N5 O5
Molecular Weight (g/mol) 517.542
Synonym (1R,2S,5S)-N-[(2S)-1-Amino-1-Oxo-3-((3S)-2-Oxopyrrolidin-3-Yl)Propan-2-Yl]-6,6-Dimethyl-3-[3-Methyl-N-(Trifluoroacetyl)-L-Valyl]-3-Azabicyclo[3.1.0]Hexane-2-Carboxamide
IUPAC Name (1R,2S,5S)-N-[(2S)-1-amino-1-oxo-3-[(3S)-2-oxopyrrolidin-3-yl]propan-2-yl]-3-[(2S)-3,3-dimethyl-2-[(2,2,2-trifluoroacetyl)amino]butanoyl]-6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxamide
SMILES CC(C)(C)[C@H](NC(=O)C(F)(F)F)C(=O)N1C[C@H]2[C@@H]([C@H]1C(=O)N[C@@H](C[C@@H]3CCNC3=O)C(=O)N)C2(C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C23H34F3N5O5/c1-21(2,3)15(30-20(36)23(24,25)26)19(35)31-9-11-13(22(11,4)5)14(31)18(34)29-12(16(27)32)8-10-6-7-28-17(10)33/h10-15H,6-9H2,1-5H3,(H2,27,32)(H,28,33)(H,29,34)(H,30,36)/t10-,11-,12-,13-,14-,15+/m0/s1
Formula Weight 517.251
Chemical Name or Material Descyano Nirmatrelvir Acetamide
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.