missing translation for 'onlineSavingsMsg'
Learn More

Descyano Citalopram Hydrobromide, TRC

Product Code. 30389177
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389177 50 mg
30403485 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389177 Supplier Toronto Research Chemicals Supplier No. TRCF59624050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1332724-07-8
Molecular Formula C19 H22 F N O . Br H
Molecular Weight (g/mol) 380.29
Synonym 1-(4-Fluorophenyl)-1,3-dihydro-N,N-dimethyl-1-isobenzofuranpropanamine Hydrobromide, Descyano Citalopram Hydrobromide
IUPAC Name 3-[1-(4-fluorophenyl)-3H-2-benzofuran-1-yl]-N,N-dimethylpropan-1-amine Hydrobromide
SMILES FC(C=C1)=CC=C1[C@@]2(CCCN(C)C)C3=CC=CC=C3CO2.Br

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H22FNO.BrH/c1-21(2)13-5-12-19(16-8-10-17(20)11-9-16)18-7-4-3-6-15(18)14-22-19;/h3-4,6-11H,5,12-14H2,1-2H3;1H
Formula Weight 379.0947
Chemical Name or Material Descyano Citalopram Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.