missing translation for 'onlineSavingsMsg'
Learn More

Desacetyl Famciclovir, TRC

Product Code. 17951345
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17951345 10 mg 10mg
17902865 100 mg 100mg
17913265 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17951345 Supplier Toronto Research Chemicals Supplier No. TRCD28872510MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 104227-88-5
Molecular Formula C12 H17 N5 O3
Molecular Weight (g/mol) 279.3
Synonym 1,3-Propanediol, 2-[2-(2-amino-9H-purin-9-yl)ethyl]-, 1-acetate, 1,3-Propanediol, 2-[2-(2-amino-9H-purin-9-yl)ethyl]-, monoacetate (ester) (9CI), BRL 43594, 2-Amino-9H-purin-9-yl)-2-(hydroxymethyl)butyl acetate, Famciclovir RC B (USP), 9-(4-Acetoxy-3-(hydroxymethyl)butyl)-2-amino-9H-purine
IUPAC Name [4-(2-aminopurin-9-yl)-2-(hydroxymethyl)butyl] acetate
SMILES CC(=O)OCC(CO)CCn1cnc2cnc(N)nc12

Specifikationer

Quantity 10 mg
InChI Formula InChI=1S/C12H17N5O3/c1-8(19)20-6-9(5-18)2-3-17-7-15-10-4-14-12(13)16-11(10)17/h4,7,9,18H,2-3,5-6H2,1H3,(H2,13,14,16)
Formula Weight 279.1331
Percent Purity >95%
Chemical Name or Material Desacetyl Famciclovir
Grade HPLC
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.