missing translation for 'onlineSavingsMsg'
Learn More

Desacetyl Cephapirin Sodium Salt, TRC

Product Code. 17971055
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17971055 25 mg 25mg
17546175 10 mg 10mg
17903125 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17971055 Supplier Toronto Research Chemicals Supplier No. TRCD28897025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104557-24-6
Molecular Formula C15 H14 N3 O5 S2 . Na
Molecular Weight (g/mol) 403.41
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-(hydroxymethyl)-8-oxo-7-[[(4-pyridinylthio)acetyl]amino]-, monosodium salt, (6R-trans)- (9CI), (6R,7R)-3-(Hydroxymethyl)-8-oxo-7-[[2-(4-pyridinylthio)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid sodium salt, Deacetylcephapirin sodium salt, Desacetylcephapirin sodium, Cefapirin-desacetyl sodium
IUPAC Name sodium;(6R,7R)-3-(hydroxymethyl)-8-oxo-7-[(2-pyridin-4-ylsulfanylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES [Na+].OCC1=C(N2[C@H](SC1)[C@H](NC(=O)CSc3ccncc3)C2=O)C(=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H15N3O5S2.Na/c19-5-8-6-25-14-11(13(21)18(14)12(8)15(22)23)17-10(20)7-24-9-1-3-16-4-2-9;/h1-4,11,14,19H,5-7H2,(H,17,20)(H,22,23);/q;+1/p-1/t11-,14-;/m1./s1
Formula Weight 403.0273
Percent Purity >95%
Chemical Name or Material Desacetyl Cephapirin Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.