missing translation for 'onlineSavingsMsg'
Learn More

Des O-isopropyl Bisoprolol, TRC

Product Code. 30366142
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366142 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30366142 Supplier Toronto Research Chemicals Supplier No. TRCD29323510MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2RS)-1-[4-((2-Hydroxyethoxy)methyl)phenoxy]-3-(isopropylamino)-2-propanol
CAS 109791-18-6
Molecular Formula C15H25NO4
Quantity 10 mg
Synonym (2RS)-1-[4-((2-Hydroxyethoxy)methyl)phenoxy]-3-(isopropylamino)-2-propanol
InChI Formula InChI=1S/C15H25NO4/c1-12(2)16-9-14(18)11-20-15-5-3-13(4-6-15)10-19-8-7-17/h3-6,12,14,16-18H,7-11H2,1-2H3
SMILES CC(C)NCC(O)COc1ccc(COCCO)cc1
IUPAC Name 1-[4-(2-hydroxyethoxymethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
Molecular Weight (g/mol) 283.3633
Formula Weight 283.1784
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.