missing translation for 'onlineSavingsMsg'
Learn More

Des(isopropylthiazolyl) Hydantoin Ritonavir (>90%), TRC

Product Code. 30362341
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362341 5 mg 5mg
30358831 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30362341 Supplier Toronto Research Chemicals Supplier No. TRCD2902155MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1010809-61-6
Molecular Formula C29 H34 N4 O5 S
Molecular Weight (g/mol) 550.67
Synonym Carbamic acid, N-[(1S,2S,4S)-2-hydroxy-4-[(4S)-4-(1-methylethyl)-2,5-dioxo-1-imidazolidinyl]-5-phenyl-1-(phenylmethyl)pentyl]-, 5-thiazolylmethyl ester, 1,3-Thiazol-5-ylmethyl N-[(1S,2S,4S)-1-benzyl-2-hydroxy-4-((4S)-4-isopropyl-2,5-dioxoimidazolidin-1-yl)-5-phenylpentyl]carbamate, Ritonavir Imp. F (EP)
IUPAC Name 1,3-thiazol-5-ylmethyl N-[(2S,3S,5S)-5-[(4S)-2,5-dioxo-4-propan-2-ylimidazolidin-1-yl]-3-hydroxy-1,6-diphenylhexan-2-yl]carbamate
SMILES CC(C)[C@@H]1NC(=O)N([C@H](C[C@H](O)[C@H](Cc2ccccc2)NC(=O)OCc3cncs3)Cc4ccccc4)C1=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C29H34N4O5S/c1-19(2)26-27(35)33(28(36)32-26)22(13-20-9-5-3-6-10-20)15-25(34)24(14-21-11-7-4-8-12-21)31-29(37)38-17-23-16-30-18-39-23/h3-12,16,18-19,22,24-26,34H,13-15,17H2,1-2H3,(H,31,37)(H,32,36)/t22-,24-,25-,26-/m0/s1
Formula Weight 550.225
Chemical Name or Material Des(isopropylthiazolyl) Hydantoin Ritonavir
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.