missing translation for 'onlineSavingsMsg'
Learn More

Des(isopropylthiazolyl) Hydantoin Ritonavir (>90%), TRC

Product Code. 30358831
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358831 50 mg 50mg
30362341 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30358831 Supplier Toronto Research Chemicals Supplier No. TRCD29021550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1010809-61-6
Molecular Formula C29 H34 N4 O5 S
Molecular Weight (g/mol) 550.67
Synonym Carbamic acid, N-[(1S,2S,4S)-2-hydroxy-4-[(4S)-4-(1-methylethyl)-2,5-dioxo-1-imidazolidinyl]-5-phenyl-1-(phenylmethyl)pentyl]-, 5-thiazolylmethyl ester, 1,3-Thiazol-5-ylmethyl N-[(1S,2S,4S)-1-benzyl-2-hydroxy-4-((4S)-4-isopropyl-2,5-dioxoimidazolidin-1-yl)-5-phenylpentyl]carbamate, Ritonavir Imp. F (EP)
IUPAC Name 1,3-thiazol-5-ylmethyl N-[(2S,3S,5S)-5-[(4S)-2,5-dioxo-4-propan-2-ylimidazolidin-1-yl]-3-hydroxy-1,6-diphenylhexan-2-yl]carbamate
SMILES CC(C)[C@@H]1NC(=O)N([C@H](C[C@H](O)[C@H](Cc2ccccc2)NC(=O)OCc3cncs3)Cc4ccccc4)C1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C29H34N4O5S/c1-19(2)26-27(35)33(28(36)32-26)22(13-20-9-5-3-6-10-20)15-25(34)24(14-21-11-7-4-8-12-21)31-29(37)38-17-23-16-30-18-39-23/h3-12,16,18-19,22,24-26,34H,13-15,17H2,1-2H3,(H,31,37)(H,32,36)/t22-,24-,25-,26-/m0/s1
Formula Weight 550.225
Chemical Name or Material Des(isopropylthiazolyl) Hydantoin Ritonavir
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.