missing translation for 'onlineSavingsMsg'
Learn More

Des-acetyl Apremilast, TRC

Product Code. 30356813
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30356813 10 mg 10mg
30356007 25 mg 25mg
30351466 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30356813 Supplier Toronto Research Chemicals Supplier No. TRCD28939810MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635705-72-5
Molecular Formula C20H22N2O6S
Molecular Weight (g/mol) 418.46
Synonym 4-Amino-2-[(1S)-1-(3-ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethyl]-1H-isoindole-1,3(2H)-dione
SMILES CCOc1cc(ccc1OC)[C@@H](CS(=O)(=O)C)N2C(=O)c3cccc(N)c3C2=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H22N2O6S/c1-4-28-17-10-12(8-9-16(17)27-2)15(11-29(3,25)26)22-19(23)13-6-5-7-14(21)18(13)20(22)24/h5-10,15H,4,11,21H2,1-3H3/t15-/m1/s1
Formula Weight 418.1199
Chemical Name or Material Des-acetyl Apremilast
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.