missing translation for 'onlineSavingsMsg'
Learn More

Des-4-hydroxy-4-nitrobenzoate Digoxigenin Monodigitoxoside, TRC

Product Code. 30382980
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382980 2.5 mg 2.5mg
30383493 10 mg 10mg
30371891 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382980 Supplier Toronto Research Chemicals Supplier No. TRCD4465142.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Des-4-hydroxy-4-nitrobenzoate Digoxigenin Monodigitoxoside
Analyte or Component Names Des-4-hydroxy-4-nitrobenzoate Digoxigenin Monodigitoxoside
Shipping Condition Room Temperature
Molecular Formula C36 H47 N O11
Synonym (2R,3R,4R,6R)-6-(((3S,5R,8R,9S,10S,12R,13S,14S,17R)-12,14-Dihydroxy-10,13-dimethyl-17-(5-oxo-2,5-dihydrofuran-3-yl)hexadecahydro-1H-cyclopenta[a]phenanthren-3-yl)oxy)-3-hydroxy-2-methyltetrahydro-2H-pyran-4-yl 4-nitrobenzoate
InChI Formula InChI=1S/C36H47NO11/c1-19-32(40)28(48-33(41)20-4-7-23(8-5-20)37(43)44)17-31(46-19)47-24-10-12-34(2)22(15-24)6-9-26-27(34)16-29(38)35(3)25(11-13-36(26,35)42)21-14-30(39)45-18-21/h4-5,7-8,14,19,22,24-29,31-32,38,40,42H,6,9-13,15-18H2,1-3H3/t19-,22-,24+,25-,26-,27+,28-,29-,31+,32-,34+,35+,36+/m1/s1
Solution Type Neat
SMILES C[C@H]1O[C@H](C[C@@H](OC(=O)c2ccc(cc2)[N+](=O)[O-])[C@@H]1O)O[C@H]3CC[C@@]4(C)[C@H](CC[C@@H]5[C@@H]4C[C@@H](O)[C@]6(C)[C@H](CC[C@]56O)C7=CC(=O)OC7)C3
IUPAC Name [(2R,3R,4R,6R)-6-[[(3S,5R,8R,9S,10S,12R,13S,14S,17R)-12,14-dihydroxy-10,13-dimethyl-17-(5-oxo-2H-furan-3-yl)-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-yl]oxy]-3-hydroxy-2-methyloxan-4-yl] 4-nitrobenzoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 669.759
Quantity 2.5 mg
Formula Weight 669.315
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.