missing translation for 'onlineSavingsMsg'
Learn More

Des-(2-propyl-1,3-dioxolane)-2-hydroxy Budesonide, TRC

Product Code. 30398876
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398876 25 mg 25mg
30391163 50 mg 50mg
30394837 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398876 Supplier Toronto Research Chemicals Supplier No. TRCH82582525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (8S,9S,10R,13S,14S)-17-(2,2-Dihydroxyacetyl)-11-hydroxy-10,13-dimethyl-6,7,8,9,10,11,12,13,14,15-decahydro-3H-cyclopenta[a]phenanthren-3-one
Analyte or Component Names (8S,9S,10R,13S,14S)-17-(2,2-Dihydroxyacetyl)-11-hydroxy-10,13-dimethyl-6,7,8,9,10,11,12,13,14,15-decahydro-3H-cyclopenta[a]phenanthren-3-one
Shipping Condition Room Temperature
Molecular Formula C21 H26 O5
Synonym 16,17-Dehydro-21-hydroxy-prednisolone
InChI Formula InChI=1S/C21H26O5/c1-20-8-7-12(22)9-11(20)3-4-13-14-5-6-15(18(24)19(25)26)21(14,2)10-16(23)17(13)20/h6-9,13-14,16-17,19,23,25-26H,3-5,10H2,1-2H3/t13-,14-,16?,17+,20-,21-/m0/s1
Solution Type Neat
SMILES C[C@]12CC(O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC=C2C(=O)C(O)O
IUPAC Name (8S,9S,10R,13S,14S)-17-(2,2-Dihydroxyacetyl)-11-hydroxy-10,13-dimethyl-6,7,8,9,10,11,12,13,14,15-decahydro-3H-cyclopenta[a]phenanthren-3-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 358.43
Quantity 25 mg
Formula Weight 358.178
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.