missing translation for 'onlineSavingsMsg'
Learn More

Des[2’-(1H-tetrazol-5-yl)] 2-Cyanolosartan, TRC

Product Code. 30354112
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354112 500 mg 500mg
30350699 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30354112 Supplier Toronto Research Chemicals Supplier No. TRCD297670500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114772-55-3
Molecular Formula C22 H22 Cl N3 O
Molecular Weight (g/mol) 379.88
Synonym 4'-[[2-butyl-4-chloro-5-(hydroxymethyl)-1H-imidazol-1-yl]methyl]- [1,1'-Biphenyl]-2-carbonitrile, 2-Butyl-4-chloro-1-[(2'-cyanobiphenyl-4-yl)methyl]-5-hydroxymethylimidazole, 4'-(2-Butyl-4-chloro-5-hydroxymethyl-1H-imidazol-1-yl)-1,1'-biphenyl-2-carbonitrile
IUPAC Name 2-[4-[[2-butyl-4-chloro-5-(hydroxymethyl)imidazol-1-yl]methyl]phenyl]benzonitrile
SMILES CCCCc1nc(Cl)c(CO)n1Cc2ccc(cc2)c3ccccc3C#N

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C22H22ClN3O/c1-2-3-8-21-25-22(23)20(15-27)26(21)14-16-9-11-17(12-10-16)19-7-5-4-6-18(19)13-24/h4-7,9-12,27H,2-3,8,14-15H2,1H3
Formula Weight 379.1451
Chemical Name or Material Des[2'-(1H-tetrazol-5-yl)] 2-Cyanolosartan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.