missing translation for 'onlineSavingsMsg'
Learn More

Des(1-azepanyl)ethyl Bazedoxifene, TRC

Product Code. 30134634
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134634 100 mg 100mg
30011569 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134634 Supplier Toronto Research Chemicals Supplier No. TRCD289720100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104599-10-2
Molecular Formula C22H19NO3
Molecular Weight (g/mol) 345.39
Synonym 2-(4-Hydroxyphenyl)-1-[(4-hydroxyphenyl)methyl]-3-methyl-1H-Indol-5-ol, 1-(4-Hydroxybenzyl)-2-(4-hydroxyphenyl)-3-methyl-1H-indol-5-ol
SMILES OC(C=C1)=CC=C1C2=C(C)C3=CC(O)=CC=C3N2CC4=CC=C(O)C=C4

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H19NO3/c1-14-20-12-19(26)10-11-21(20)23(13-15-2-6-17(24)7-3-15)22(14)16-4-8-18(25)9-5-16/h2-12,24-26H,13H2,1H3
Formula Weight 345.1365
Chemical Name or Material Des(1-azepanyl)ethyl Bazedoxifene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.