missing translation for 'onlineSavingsMsg'
Learn More

DERP13 Antibody (2B11), Novus Biologicals™

Product Code. 18419389 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419389 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419389 Supplier Novus Biologicals Supplier No. H00079783M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DERP13 Monoclonal antibody specifically detects DERP13 in Human samples. It is validated for ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen DERP13
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2B11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_079004
Gene Alias C7orf10, CaiB/baiF CoA-transferase family protein C7orf10, chromosome 7 open reading frame 10, Dermal papilla-derived protein 13, DERP13, DERP13FLJ11808, EC 2.-, FLJ53872, ORF19, Russel-Silver syndrome candidate
Host Species Mouse
Immunogen C7orf10 (NP_079004.1, 335 a.a. ∽ 434 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AVSGFQSLLHSLAHGPFLHLQGSARVLHNGLVMEMEHPTVGKISVPGPAVRYSKFKMSEARPPPLLGQHTTHILKEVLRYDDRAIGELLSAGVVDQHETH
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79783
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.