missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dermokine beta Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92134-0.02ml
This item is not returnable.
View return policy
Description
Dermokine beta Polyclonal antibody specifically detects Dermokine beta in Human, Rat samples. It is validated for Western Blot
Specifications
| Dermokine beta | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| dermokine, Epidermis-specific secreted protein SK30/SK89, FLJ79374, UNQ729, ZD52F10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 355-449 of human DMKN (NP_001177276.1). WEDFKQNTPFLNWKAIIEGADASSLQKRAGRADQPGAGWQEVAAVTSKNYNYNQHAYPTAYGGKYSVKTPAKGGVSPSSSASRVQPGLLQWVKFW | |
| 0.02 mL | |
| Cell Biology | |
| 93099 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction