missing translation for 'onlineSavingsMsg'
Learn More

Dermofungin, TRC

Product Code. 30133713
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133713 50 g 50g
30136156 10 g 10g
30007228 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133713 Supplier Toronto Research Chemicals Supplier No. TRCD28193050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-16-5
Molecular Formula C9 H6 Cl N O
Molecular Weight (g/mol) 179.6
Synonym Cloxiquine, 5-Chloro-8-hydroxyquinoline, 5-Chloro-8-oxyquinoline, 5-Chloro-8-quinolinol, 5-Chloroquinoline-8-ol, 5-Chlorooxine, 8-Hydroxy-5-chloroquinoline, Cloxyquin, Dermofongin, Dermofungin, NSC 35083, NSC 53182, NSC 74943, Clioquinol Imp. A (EP)
IUPAC Name 5-chloroquinolin-8-ol
SMILES Oc1ccc(Cl)c2cccnc12

Specifications

Quantity 50 g
InChI Formula InChI=1S/C9H6ClNO/c10-7-3-4-8(12)9-6(7)2-1-5-11-9/h1-5,12H
Formula Weight 179.0138
Percent Purity >95%
Chemical Name or Material Cloxiquine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.