missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dermatopontin Mouse anti-Human, Rat, Clone: 4B1A12, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | Dermatopontin |
|---|---|
| Clone | 4B1A12 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16808145
|
Proteintech
66348-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898135
|
Proteintech
66348-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dermatopontin is an extracellular matrix protein with possible functions in cell-matrix interactions and matrix assembly. The protein is found in various tissues and many of its tyrosine residues are sulphated. Dermatopontin is postulated to modify the behavior of TGF-beta through interaction with decorin.Specifications
| Dermatopontin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| dermatopontin, DPT, TRAMP | |
| Dpt | |
| IgG2b | |
| Protein A | |
| Antibody |
| 4B1A12 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| Q07507 | |
| 1805, 289178 | |
| Dermatopontin Fusion Protein Ag6200 | |
| Primary | |
| -20°C | |
| DPT |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title