missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Derlin 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97369-50ul
147.05 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Derlin 1 Polyclonal antibody specifically detects Derlin 1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Derlin 1 | |
| Polyclonal | |
| Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| DER-1, DER1Degradation in endoplasmic reticulum protein 1, Der1-like domain family, member 1, Der1-like protein 1, derlin-1, DERtrin-1, FLJ13784, FLJ42092, MGC3067, PRO2577 | |
| Produced in rabbits immunized with E. coli-derived Human Derlin 1 fragment. | |
| 50 μg | |
| ER Markers, Immunology, Lipid and Metabolism, Protein Turnover, Signal Transduction, Unfolded Protein Response | |
| 79139 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction