missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DERL1, Mouse anti-Human, Clone: 1B9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00079139-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: AQLNRDMIVSFWFGTRFKACYLPWVILGFNYIIGGSVINELIGNLVGHLYFFLMFRYPMDLGGRNFLSTPQFLYRWLPSRRGGVSGFGVPPASMRRAADQSpecifications
| DERL1 | |
| Monoclonal | |
| Unconjugated | |
| Der1-like domain family, member 1 | |
| DER-1/DER1/FLJ13784/FLJ42092/MGC3067/PRO2577 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1B9 | |
| In 1x PBS, pH 7.4 | |
| BC002457 | |
| DERL1 | |
| DERL1 (AAH02457, 134 a.a. ∼ 233 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 79139 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction