missing translation for 'onlineSavingsMsg'
Learn More

DEPTOR/DEPDC6 Antibody [PerCP], Novus Biologicals™

Product Code. 30734283 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734283 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734283 Supplier Novus Biologicals Supplier No. AF7255C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

DEPTOR/DEPDC6 Polyclonal antibody specifically detects DEPTOR/DEPDC6 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEPTOR/DEPDC6
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias DEP domain containing 6, DEP domain containing mTOR interacting protein, DEP domain containing MTOR-interacting protein, DEP domain-containing mTOR-interacting protein, DEP domain-containing protein 6, DEP.6, DEPDC6FLJ12341, DKFZp564B1778, FLJ12428, FLJ13854
Host Species Sheep
Immunogen E. coli-derived recombinant human DEPTOR/DEPDC6, Ala31-Gly158, Accession # Q8TB45
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64798
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.