missing translation for 'onlineSavingsMsg'
Learn More

DEPTOR/DEPDC6 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 13 publications

Brand:  Novus Biologicals NBP1-49674SS

 View more versions of this product

Product Code. 18729283

  • £196.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DEPTOR/DEPDC6 Polyclonal specifically detects DEPTOR/DEPDC6 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DEPTOR/DEPDC6
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
DEP domain containing 6, DEP domain containing mTOR interacting protein, DEP domain containing MTOR-interacting protein, DEP domain-containing mTOR-interacting protein, DEP domain-containing protein 6, DEP.6, DEPDC6FLJ12341, DKFZp564B1778, FLJ12428, FLJ13854
Rabbit
Affinity Purified
RUO
Primary
Human and mouse. Predicted to react with bovine based on 100% sequence homology.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1.0 mg/mL
Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100
Q8TB45, Q570Y9
DEPTOR
A genomic peptide made to an internal region of the human DEPTOR protein (within residues 10-200). [Swiss-Prot: Q8TB45]
0.025 mL
Cancer, Signal Transduction
64798
Human, Mouse, Rat, Bovine
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only