Learn More
Buerkle™ Depth Stop for Sampler
Product Code. 31188927 Shop All Bürkle Products
Change view
Click to view available options
Material:
Aluminum
Stainless Steel (V4A)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material
31188927 Stainless Steel (V4A)
31188867 Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31188927 Supplier Buerkle™ Supplier No. 53170015
  • ONLINE EXCLUSIVE PRICE
    £156.00
    £146.91 / Pack of 1
    Save £9.09
    5% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For a sampling procedure to be reproducible, it must always be carried out under the same conditions. The depth stop for samplers is a useful accessory for this purpose. The depth stop is attached to the sampler at any height using a C-clamp screw.
This ensures that the sampler penetrates to the same desired depth each time it is inserted. The penetration depth is thus fixed and can be precisely recorded in a test report or sampling instructions, for example. The depth stop can also prevent unintentional piercing during sampling.
  • Enables standardized and therefore reproducible sampling
  • Very good clamping effect due to C-clamping screw
  • Available for 25/50 mm diameter, suitable for many sampler lances
TRUSTED_SUSTAINABILITY
Specifications
Product Type Sampler Insert
Material Stainless Steel (V4A)
Autoclavable Autoclavable