missing translation for 'onlineSavingsMsg'
Learn More

DEPDC7 Antibody (4D9), Novus Biologicals™

Product Code. 18408999 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408999 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408999 Supplier Novus Biologicals Supplier No. H00091614M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DEPDC7 Monoclonal antibody specifically detects DEPDC7 in Human samples. It is validated for ELISA, Immunoprecipitation, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEPDC7
Applications ELISA, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 4D9
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_631899
Gene Alias DEP domain containing 7, DEP domain-containing protein 7, dJ85M6.4, dJ85M6.4 (novel 58.3 KDA protein), novel 58.3 KDA protein, Protein TR2/D15, TR2
Host Species Mouse
Immunogen LOC91614 (NP_631899, 393 a.a. ∽ 499 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NPSEFKLQKESDNRMVVKRIFSKAIVDNKNLSKGKTDLLVLFLMDHQKDVFKIPGTLHKIVSVKLMAIQNGRDPNRDAGYIYCQRIDQRDYSNNTEKTTKDELLNLL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91614
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.