missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEPDC6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21312-1-AP-150UL
This item is not returnable.
View return policy
Description
Negative regulator of the mTORC1 and mTORC2 signaling pathways. Inhibits the kinase activity of both complexes.Specifications
| DEPDC6 | |
| Polyclonal | |
| Unconjugated | |
| DEPTOR | |
| DEP domain containing 6, DEP.6, DEPDC6, Deptor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64798 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8TB45 | |
| DEPTOR | |
| DEPDC6 Fusion Protein Ag15959 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction