missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEP-1/CD148 Antibody (143-41), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals FAB1934H
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
DEP-1/CD148 Monoclonal antibody specifically detects DEP-1/CD148 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF
Especificaciones
| DEP-1/CD148 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF | |
| 143-41 | |
| Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF-ready | |
| CD148, CD148 antigen, Density-enhanced phosphatase 1, DEP-1, DEP1EC 3.1.3.48, HPTP eta, HPTPeta, human density enhanced phosphatase-1, protein tyrosine phosphatase, receptor type, J, protein tyrosine phosphatase, receptor type, J polypeptide, Protein-tyrosine phosphatase eta, Protein-tyrosine phosphatase receptor type J, receptor-type tyrosine-protein phosphatase eta, R-PTP-eta, R-PTP-J, SCC1, susceptibility to colon cancer 1, mouse, homolog of | |
| Phytohemagglutinin-stimulated human peripheral blood mononuclear cells | |
| 0.1 mL | |
| Protein Phosphatase | |
| 5795 | |
| Store at 4°C in the dark. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido