missing translation for 'onlineSavingsMsg'
Learn More

Deoxystreptamine-kanosaminide, TRC

Product Code. 30005475
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005475 50 mg 50mg
17983425 10 mg 10mg
17972025 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005475 Supplier Toronto Research Chemicals Supplier No. TRCD26351050MG
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20744-51-8
Molecular Formula C12 H25 N3 O7
Molecular Weight (g/mol) 323.34
Synonym D-Streptamine, 6-O-(3-amino-3-deoxy-α-D-glucopyranosyl)-2-deoxy-, Streptamine, 4-O-(3-amino-3-deoxy-α-D-glucopyranosyl)-2-deoxy-, L- (8CI), 6-O-(3-Amino-3-deoxy-α-D-glucopyranosyl)-2-deoxy-D-streptamine, Antibiotic NK 1012-2, Deoxystreptamine kanosaminide
IUPAC Name (2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol
SMILES N[C@H]1C[C@@H](N)[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](N)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H25N3O7/c13-3-1-4(14)11(10(20)7(3)17)22-12-9(19)6(15)8(18)5(2-16)21-12/h3-12,16-20H,1-2,13-15H2/t3-,4+,5+,6-,7+,8+,9+,10-,11-,12+/m0/s1
Formula Weight 323.1693
Percent Purity >95%
Chemical Name or Material Deoxystreptamine-kanosaminide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.