missing translation for 'onlineSavingsMsg'
Learn More

Deoxyribonucleic Acid Sodium Salt, Fisher BioReagents

Product Code. 10428923
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
Packaging:
Amber Glass
Unit Size:
10g
1g
250mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10428923 5 g Amber Glass 5g
10502955 250 mg Amber Glass 250mg
11403074 1 g Amber Glass 1g
11361126 10 g Amber Glass 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10428923 Supplier Fisher BioReagents Supplier No. BP25145
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Deoxyribonucleic Acid Sodium Salt is used in several biochemical research applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68938-01-2
Molecular Formula C17H15BrFNO3
Molecular Weight (g/mol) 50000-100000
MDL Number MFCD00130922
InChI Key GXZCYECDYOBPGH-UHFFFAOYNA-N
PubChem CID 73904662
IUPAC Name (2R)-3-(3-bromophenyl)-2-[[2-(4-fluorophenyl)acetyl]amino]propanoic acid
SMILES *

Specifications

Color White
Loss on Drying 14% max.
Packaging Amber Glass
Quantity 5 g
Formula Weight 6000000
Physical Form Solid
Chemical Name or Material Deoxyribonucleic Acid Sodium Salt, From Salmon Sperm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.