missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Deoxyribonuclease I (Bovine)
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
#REF!
Specifications
Specifications
| Accession Number | P00639 |
| For Use With (Application) | Enzyme Activity |
| Gene ID (Entrez) | 282217 |
| Molecular Weight (g/mol) | 31.3kDa |
| Name | Deoxyribonuclease I (Bovine) |
| Preparation Method | Native protein chromatographically purified from Bovine Pancreas |
| Quantity | 5 mg |
| Storage Requirements | Store at 4°C on dry atmosphere. |
| Regulatory Status | RUO |
| Common Name | DNASE1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction