missing translation for 'onlineSavingsMsg'
Learn More

Deoxypyridinoline Chloride Trihydrochloride Salt, TRC

Product Code. 30357818
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357818 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357818 Supplier Toronto Research Chemicals Supplier No. TRCD24990010MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Deoxypyridinoline chloride trihydrochloride
CAS 204074-56-6
Molecular Formula C18 H28 N4 O7 . H Cl
Quantity 10 mg
Synonym 4-[(2S)-2-Amino-2-carboxyethyl]-1-[(5S)-5-amino-5-carboxypentyl]-3-[(3S)-3-amino-3-carboxypropyl]-5-hydroxypyridinium Chloride Trihydrochloride Salt, Lysylpyridinoline
InChI Formula InChI=1S/C18H28N4O7.ClH/c19-12(16(24)25)3-1-2-6-22-8-10(4-5-13(20)17(26)27)11(15(23)9-22)7-14(21)18(28)29;/h8-9,12-14H,1-7,19-21H2,(H3-,23,24,25,26,27,28,29);1H/t12-,13-,14-;/m0./s1
SMILES Cl.N[C@@H](CCCC[n+]1cc(O)c(C[C@H](N)C(=O)[O-])c(CC[C@H](N)C(=O)O)c1)C(=O)O
IUPAC Name (2S)-2-amino-3-[1-[(5S)-5-amino-5-carboxypentyl]-3-[(3S)-3-amino-3-carboxypropyl]-5-hydroxypyridin-1-ium-4-yl]propanoate;hydrochloride
Molecular Weight (g/mol) 448.899
Formula Weight 448.172
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.