missing translation for 'onlineSavingsMsg'
Learn More

Deoxynojirimycin Hydrochloride, TRC

Product Code. 30362717
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362717 50 mg 50mg
30354286 10 mg 10mg
30350359 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30362717 Supplier Toronto Research Chemicals Supplier No. TRCD24500550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73285-50-4
Molecular Formula C6 H13 N O4 . Cl H
Molecular Weight (g/mol) 199.63
Synonym 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride (1:1), (2R,3R,4R,5S)-, 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride, (2R,3R,4R,5S)- (9CI), 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride, [2R-(2α,3β,4α,5β)]-, (+)-1-Deoxynojirimycin hydrochloride, 1-Deoxynojirimycin hydrochloride, AT 2220, Duvoglustat hydrochloride, Moranoline hydrochloride
IUPAC Name (2R,3R,4R,5S)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
SMILES Cl.OC[C@H]1NC[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H13NO4.ClH/c8-2-3-5(10)6(11)4(9)1-7-3;/h3-11H,1-2H2;1H/t3-,4+,5-,6-;/m1./s1
Formula Weight 199.0611
Chemical Name or Material Deoxynojirimycin Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.