missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deoxycytidylate deaminase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | Deoxycytidylate deaminase |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231061
|
Novus Biologicals
NBP3-38207-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228082
|
Novus Biologicals
NBP3-38207-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Deoxycytidylate deaminase Polyclonal antibody specifically detects Deoxycytidylate deaminase in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| Deoxycytidylate deaminase | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.3), 50% glycerol | |
| 1635 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| dCMP deaminaseEC 3.5.4.12, deoxycytidylate deaminase, MGC111062 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-178 of human Deoxycytidylate deaminase (NP_001912.2).,, Sequence:, MSEVSCKKRDDYLEWPEYFMAVAFLSAQRSKDPNSQVGACIVNSENKIVGIGYNGMPNGCSDDVLPWRRTAENKLDTKYPYVCHAELNAIMNKNSTDVKGCSMYVALFPCNECAKLIIQAGIKEVIFMSDKYHDSDEATAARLLFNMAGVTFRKFIPKCSKIVIDFDSINSRPSQKLQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title