missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Deoxycytidine triphosphate

Product Code. 16499876
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499876 50 mg 50mg
16443348 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499876 Supplier MedChemExpress Supplier No. HY101400

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Deoxycytidine triphosphate (dCTP) is a nucleoside triphosphate that can be used for DNA synthesis. Deoxycytidine triphosphate has many applications, such as real-time PCR, cDNA synthesis, and DNA sequencing.

Specifications

CAS 2056-98-6
Content And Storage Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Product Type Biochemical Assay Reagents
Quantity 50 mg
Shipping Condition Room temperature
Synonym dCTP ∣ 2′-Deoxycytidine-5′-triphosphate
Molecular Weight (g/mol) 467.16 g/mol
Solubility H2O : 50 mg/mL (107.03 mM; Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.