missing translation for 'onlineSavingsMsg'
Learn More

Deoxycorticosterone acetate, MedChemExpress

Product Code. 16408776
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16408776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16408776 Supplier Medchem Express Supplier No. HYB1472
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Deoxycorticosterone acetate is a steroid hormone produced by the adrenal gland that possesses mineralocorticoid activity and acts as a precursor to aldosterone.

Deoxycorticosterone acetate is a steroid hormone produced by the adrenal gland that possesses mineralocorticoid activity and acts as a precursor to aldosterone.

Specifications

Chemical Name or Material Deoxycorticosterone acetate
CAS 56-47-3
Color White
Purity Grade Notes Research
Molecular Formula C23H32O4
Quantity 100 mg
Synonym 11-Deoxycorticosterone acetate DOC acetate Cortexone acetate
Solubility Information DMSO : 41.67 mg/mL (111.87 mM; ultrasonic and warming and heat to 80°C) ∣H2O : 1 mg/mL (2.68 mM; ultrasonic and warming and heat to 80°C)
SMILES C[C@@]12[C@@H](C(COC(C)=O)=O)CC[C@@]1([H])[C@]3([H])CCC4=CC(CC[C@]4(C)[C@@]3([H])CC2)=O
Molecular Weight (g/mol) 372.5
Formula Weight 372.5
Percent Purity 98.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.