missing translation for 'onlineSavingsMsg'
Learn More

Deoxycholic acid, MedChemExpress

Product Code. 16489106
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489106 100 mg 100mg
16422386 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16489106 Supplier Medchem Express Supplier No. HYN0593
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Deoxycholic acid (cholanoic acid), a bile acid, is a by-product of intestinal metabolism, that activates the G protein-coupled bile acid receptorTGR5.

Deoxycholic acid (cholanoic acid), a bile acid, is a by-product of intestinal metabolism, that activates the G protein-coupled bile acid receptorTGR5.

Specifications

Chemical Name or Material Deoxycholic acid
CAS 83-44-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H40O4
Quantity 100 mg
Synonym Cholanoic Acid Desoxycholic acid
Solubility Information DMSO : ≥ 100 mg/mL (254.73 mM) ∣H2O : 1 mg/mL (2.55 mM; ultrasonic and warming and heat to 80°C)
SMILES C[C@@]1([C@@]2([H])[C@H](C)CCC(O)=O)[C@](CC2)([H])[C@@](CC[C@@]3([H])[C@@]4(CC[C@@H](O)C3)C)([H])[C@]4([H])C[C@@H]1O
Molecular Weight (g/mol) 392.57
Formula Weight 392.57
Percent Purity 97.21%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.