missing translation for 'onlineSavingsMsg'
Learn More

Deoxybenzoin, 97%

Product Code. 11413244
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413244 100 g 100g
11403244 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413244 Supplier Thermo Scientific Alfa Aesar Supplier No. A10580.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 451-40-1 | C14H12O | 196.25 g/mol

Deoxybenzoin, is used as a benzoin derivative and as a photoinitiator in vinyl polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Deoxybenzoin, is used as a benzoin derivative and as a photoinitiator in vinyl polymerization.

Solubility
Soluble in water (partly), methanol, alcohols, and ketones.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 451-40-1
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00003081
InChI Key OTKCEEWUXHVZQI-UHFFFAOYSA-N
Synonym 2-phenylacetophenone, deoxybenzoin, benzyl phenyl ketone, ethanone, 1,2-diphenyl, desoxybenzoin, 1,2-diphenylethan-1-one, phenyl benzyl ketone, benzoin, deoxy, acetophenone, 2-phenyl, deoxy benzoin
PubChem CID 9948
IUPAC Name 1,2-diphenylethanone
SMILES O=C(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 53°C to 57°C
Density 1.2
Boiling Point 319°C to 321°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 1072876
Solubility Information Soluble in water (partly),methanol,alcohols,and ketones.
Formula Weight 196.25
Percent Purity 97%
Chemical Name or Material Deoxybenzoin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.