missing translation for 'onlineSavingsMsg'
Learn More

Deoxyarbutin, MedChemExpress

Product Code. 16472276
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16472276

Brand: Medchem Express HYB1461

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Deoxyarbutin is a new effective lighten ingredient, can effectively inhibit tyrosinase activity and melanin synthesis to get significant and lasting lightening effect.

Deoxyarbutin is a new effective lighten ingredient, can effectively inhibit tyrosinase activity and melanin synthesis to get significant and lasting lightening effect.

Specifications

Chemical Name or Material Deoxyarbutin
CAS 53936-56-4
Color White
Purity Grade Notes Research
Molecular Formula C11H14O3
Quantity 500 mg
Solubility Information DMSO : ≥ 100 mg/mL (514.85 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC1=CC=C(OC2CCCCO2)C=C1
Molecular Weight (g/mol) 194.23
Formula Weight 194.23
Percent Purity 97.35%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.