missing translation for 'onlineSavingsMsg'
Learn More

Density Standards (15 °C), Paragon Scientific™

Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C

Brand:  Paragon Scientific DEN15-03

Additional Details : CAS Number : 64742-88-7 Weight : 0.06000kg Transport : UN number : 1268 Chem class : 3 Pack group : III


 View more versions of this product

Product Code. 13430479

  • £81.75 / 60mL
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols.
  • Supplied in tamper evident packaging with a 2 year shelf life.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • Manufactured in the United Kingdom
Specifications

Specifications

60 mL
0.8118 g/mL (15°C)
Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DEN15-03

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H372-Causes damage to organs.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Nominal values quoted