missing translation for 'onlineSavingsMsg'
Learn More

Density Standards (15 °C), Paragon Scientific™

Product Code. 13400479
Change view
Click to view available options
Density:
0.6654 g/mL (15°C)
0.7183 g/mL (15°C)
0.7807 g/mL (15°C)
0.8111 g/mL (15°C)
0.8494 g/mL (15°C)
0.8648 g/mL (15°C)
0.8811 g/mL (15°C)
0.9413 g/mL (15°C)
0.9823 g/mL (15°C)
1.0248 g/mL (15°C)
1.0687 g/mL (15°C)
1.1280 g/mL (15°C)
1.1962 g/mL (15°C)
1.2829 g/mL (15°C)
1.6300 g/mL (15°C)
Unit Size:
60mL
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Density unitSize
13400479 0.6654 g/mL (15°C) 60mL
13440479 0.7183 g/mL (15°C) 60mL
13420479 0.8111 g/mL (15°C) 60mL
13430479 0.7807 g/mL (15°C) 60mL
13410479 0.8494 g/mL (15°C) 60mL
13470479 0.8648 g/mL (15°C) 60mL
13450479 0.8811 g/mL (15°C) 60mL
13460479 0.9413 g/mL (15°C) 60mL
13480479 0.9823 g/mL (15°C) 60mL
13490479 1.0248 g/mL (15°C) 60mL
13410489 1.0687 g/mL (15°C) 60mL
13420489 1.1280 g/mL (15°C) 60mL
13430489 1.1962 g/mL (15°C) 60mL
13440489 1.2829 g/mL (15°C) 60mL
13460489 1.6300 g/mL (15°C) 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 13400479 Supplier Paragon Scientific Supplier No. DEN1501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C

  • Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols.
  • Supplied in tamper evident packaging with a 2 year shelf life.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • Manufactured in the United Kingdom

Specifications

Density 0.6654 g/mL (15°C)
Certifications/Compliance ISO 17025, UKAS ISO 17034
Quantity 60 mL
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • DEN15-01
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Reproductive toxicity Category 2
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P273-Avoid release to the environment.

Nominal values quoted

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.