missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Density reference standard, density reference material
- Tested by vibrational densitometer and can be used as a calibration or control standard for density measurement by vibrational or hydrometer (specific gravity) based techniques.
- Reagecon holds ISO 17025 Accreditation for testing density standards using vibrational densitometers (INAB Ref: 264T).
- The product is prepared gravimetrically on a weight/weight basis.
- Both solute and solvent are weighed on a balance calibrated by OIML traceable weights.
- Reagecon holds ISO 17025 Accreditation for calibration of laboratory balances (INAB Ref: 265C).
- The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
- The density of the product is established and certified using high performance calibrated vibrational densitometers.
- High accuracy (± 0.001642 g/mL)
Specifications
Specifications
| Packaging | Amber glass Winchester bottle |
| Physical Form | Liquid |
| Concentration | 1.0265 g/mL at 25°C (±0.001642) |
| Quantity | 100 mL |
Safety and Handling
Product Identifier
- density standard 1.0265g/ml @ 25
- Danger
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H335-May cause respiratory irritation.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P361+P364-Take off immediately all contaminated clothing and wash it before reuse.
- P321-Specific treatment (see on this label).
- P330-Rinse mouth.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: pentanol isomers
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
