missing translation for 'onlineSavingsMsg'
Learn More

Density Standard Quality Range 0.9969 g/mL at 25°C, Reagecon
BIOPHARMA

Product Code. 12802553 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12802553 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12802553 Supplier Reagecon Supplier No. DEN25140

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Manufactured and tested in accordance with ASTM D4052 for testing of density or relative density (specific and API gravity).

  • Density reference standard, density reference material
  • Tested by vibrational densitometer and can be used as a calibration or control standard for density measurement by vibrational or hydrometer (specific gravity) based techniques.
  • Reagecon holds ISO 17025 Accreditation for testing density standards using vibrational densitometers (INAB Ref: 264T).
  • The product is prepared gravimetrically on a weight/weight basis.
  • Both solute and solvent are weighed on a balance calibrated by OIML traceable weights.
  • Reagecon holds ISO 17025 Accreditation for calibration of laboratory balances (INAB Ref: 265C).
  • The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
  • The density of the product is established and certified using high performance calibrated vibrational densitometers.
  • High accuracy (± 0.001642 g/mL)

Specifications

Packaging Amber glass Winchester bottle
Physical Form Liquid
Concentration 0.9969 g/mL at 25°C (±0.001642)
Quantity 100 mL
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 2
  • Acute toxicity Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H300 + H310-Fatal if swallowed or in contact with skin.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P331-Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contain: Poly(ethylene glycol), pentanol isomers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.