Learn More
Density Standard (15°C); UKAS ISO/IEC17025 and ISO 17034 certified, nominal density 0.6643 g/mL (15°C), 500 mL, Paragon Scientific™
Product Code. 16020212
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 16020212
Brand: Paragon Scientific DEN1501500ML
Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C to 150°C
  • Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols
  • Supplied in tamper evident packaging with a 2 year shelf life
  • Manufactured in the United Kingdom
Specifications
UN Number UN 1208
Quantity 500 mL
Nominal values quoted. Certified Values will vary from batch to batch.