missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ Density Kit, XPR/XSR-Analytical
Shop All Mettler Toledo Products
Click to view available options
:
Density Kit, XPR/XSR-Analytical
Description
- Density Kits are easily installed on Excellence Balances for fast and precise density determination of solid, liquid, porous and pasty substances
- Fully integrated workflows for five different methods: The built-in density application guides you step-by-step through the procedure
- The application software collects the density values, calculates the results and allows statistical evaluation of the measurement series
Specifications
Specifications
| For Use With (Application) | Measuring density |
| Type | Density Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction